VWR®, Plain Microscope Slides
Supplier: VWR Collection
VWRI631-1160EA 112 SGD
VWRI631-1160
VWR®, Plain Microscope Slides
Slides Microscope Slides
Microscope slides with ground edges designed for human blood or fluid tests.
  • Shelf life of 60 months
  • Smooth edges for operational safety
Suitable for use in haematology and microbiology applications.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: Fragile items should be stored in dry and proper laboratory temperature conditions.