VWR®, Double-Frosted Microscope Slides
Supplier: VWR Collection
VWRI631-1161EA 169 SGD
VWRI631-1161
VWR®, Double-Frosted Microscope Slides
Slides Microscope Slides
Microscope slides with double-frosted ends designed for human blood, body fluids tests, as well as paraffin sections.
  • Shelf life of 60 months
  • Smooth edges for operational safety
  • Possible marking with pencils, marker pens and labelling systems
Suitable for use in haematology, microbiology, as well as histopathology applications.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: Fragile items should be stored in dry and proper laboratory temperature conditions.
Order Now
SPECIFICATIONS
Environmentally Preferable