VWR®, Silane Coated, Colour Frosted Slides
Supplier: VWR Collection
VWRI631-1163EA 447 SGD
VWRI631-1163 VWRI631-1164 VWRI631-1165 VWRI631-1166 VWRI631-1167 VWRI631-1168
VWR®, Silane Coated, Colour Frosted Slides
Slides Adhesive Slides
Silane coated (aminoalkylsilane) glass slides producing a sticky surface, which can bind tissue sections to glass.
  • Smooth edges for operational safety
  • Marking with pencils, marker pens and conventional labelling systems
  • Shelf life of 18 months
Recommended for H&E stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: Fragile items should be stored in dry and proper laboratory temperature conditions. Not tested for use in microarray assays.