VWR®, Inkjet Printer Slides
Supplier: VWR Collection
VWRI631-1177EA 8980 SGD
VWRI631-1177 VWRI631-1178 VWRI631-1179 VWRI631-1182
VWR®, Inkjet Printer Slides
Slides Microscope Slides
Microscopy slides are designed with colour-frosted ends for H&E stain on frozen sections in histopathology examination.
  • Shelf life of 60 months
  • Pre-cleaned, without mark, print or contamination risks
  • Marking with conventional labelling system, pencil, marker pens and inkjet printers
Specially produced for all inkjet printers.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: Fragile items should be stored in dry and proper laboratory temperature conditions.
Order Now
SPECIFICATIONS
Environmentally Preferable