VWR®, Colour-Frosted Microscope Slides
Supplier: VWR Collection
VWRI631-1554EA 7.6 SGD
VWRI631-1554 VWRI631-1555 VWRI631-1556 VWRI631-1557 VWRI631-1558 VWRI631-1559 VWRI631-1560 VWRI631-1561 VWRI631-1562 VWRI631-1563 VWRI631-1564 VWRI631-1565
VWR®, Colour-Frosted Microscope Slides
Slides Microscope Slides
Frosted ends in six different colours.
  • Available with either cut or bevelled edges
  • Writing surface resistant to all common dyes and solvents
W×D×H: 76×26×1 mm
Order Now
SPECIFICATIONS
Environmentally Preferable