VWR®, Macro Cuvettes
Supplier: VWR Collection
VWRI634-1067EA 29.1 SGD
VWRI634-1067
VWR®, Macro Cuvettes
Cuvettes Spectrophotometer Cuvettes
Disposable macro cuvettes, four clear sides.
  • Clear and clean optical path
  • Four windows for precise measurements
  • Manufactured under controlled room conditions
Specially designed for fluorescence spectroscopy.