VWR®, Absorption Cuvettes
Supplier: VWR Collection
VWRI634-1196EA 246 SGD
VWRI634-1196 VWRI634-1197 VWRI634-1198 VWRI634-1199 VWRI634-1200 VWRI634-1201 VWRI634-1202 VWRI634-1203 VWRI634-1204 VWRI634-1205 VWRI634-1206 VWRI634-1207 VWRI634-1208 VWRI634-1209 VWRI634-1210 VWRI634-1211 VWRI634-1212 VWRI634-1213 VWRI634-1214 VWRI634-1215 VWRI634-1216 VWRI634-1217 VWRI634-1218 VWRI634-1219 VWRI634-1220 VWRI634-1221 VWRI634-1222
VWR®, Absorption Cuvettes
Cuvettes Spectrophotometer Cuvettes
High quality cuvettes manufactured from quartz or optical glass for accurate spectrophotometric analysis.
  • Fully heat-fused construction
  • Two cover types available: Flat lid and stopper
  • Matched sets available
Due to the fully heat-fused construction (no intermediate materials or adhesives), and careful annealing to remove any strain, there is no optical or physical distortion of the two optically polished windows. Flat lids and stoppers are made of polytetrafluoroethylene (PTFE). Stoppered cells provide a more suitable seal for volatile liquids. MatchedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable