VWR® PV4 Visible Spectrophotometer
Supplier: VWR Collection
VWRI634-1228EA 2650 SGD
VWRI634-1228
VWR® PV4 Visible Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The PV4 visible spectrophotometer is an easy to use instrument with advanced performance.
Highly accurate and reliable, with several basic functions, such as photometry test, quantitation test and spectrum scanning.
Delivery information: Delivered with spectrophotometer, glass cuvettes (4 pcs.), power cord (Euro/UK/CH plug), software USB drive (1 pc.), USB cable (1 pc.), quick manual (1 pc.), dust cover (1 pc.), fuse (1 pc.).