VWR® P6, UV/Visible Spectrophotometer
Supplier: VWR Collection
VWRI634-1230EA 5910 SGD
VWRI634-1230
VWR® P6, UV/Visible Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The P6 UV/Visible single beam spectrophotometer is an easy to use instrument with advanced performance.
  • Highly accurate and reliable
  • Simple, elegant design with aluminum base
  • Wireless tablet control
  • Easy to change lamp
  • User friendly interface
  • 64 GB internal storage capabilites
  • USB-C, WI-FI, Bluetooth, HDMI connectivity
Provides several basic functions, such as photometry test, multi wavelength scanning, kinetics, quantitation test, spectrum scanning and biological methods.
The instrument is shipped with VWR® UV studio standard software, sampleMore Product Information