VWR® P9, UV/Visible Spectrophotometer
Supplier: VWR Collection
VWRI634-1231EA 9020 SGD
VWRI634-1231
VWR® P9, UV/Visible Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The P9 UV/Visible double-beam spectrophotometer is an easy to use instrument with advanced performance.
  • Highly accurate and reliable
  • Simple, elegant design with aluminum base
  • Easy to change lamp
  • User friendly interface
  • 64 GB internal storage capabilites
  • USB-A + B, Ethernet, VGA, HDMI connectivity
Provides several basic functions, such as photometry test, multi wavelength scanning, kinetics, quantitation test, spectrum scanning and biological methods.
The instrument is shipped with VWR® UV studio standard software, sample cuvettes (glass/quartz),More Product Information