VWR® M9 UV/Visible Spectrophotometer
Supplier: VWR Collection
VWRI634-1237EA 11150 SGD
VWRI634-1237
VWR® M9 UV/Visible Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The M9 UV/Vis Spectrophotometer is a double beam UV/Vis spectrophotometer with a variable spectral bandwidth for routine and advanced applications.
  • Double beam optical system providing a high-performance and high-precision experience
  • Xenon flash lamp
  • Variable spectral bandwidth 0,5 nm; 1 nm; 2 nm; 4 nm; 5 nm
  • Built-in 10.1" IPS capacitive color touch screen, no external computer required
  • 64 GB internal storage capacity
  • USB-A, USB-B, Ethernet, VGA, HDMI connectivity
  • Extensive range of accessories for microvolume measurement, optical fibre accessory, sipper
More Product Information