VWR® UV-3100PC, UV/Visible Spectrophotometer
Supplier: VWR Collection
VWRI634-6042EA 5820 SGD
VWRI634-6042
VWR® UV-3100PC, UV/Visible Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
UV/Visible spectrophotometer for advanced measurements in quality control, biochemical and pharmaceutical applications.
  • Absorption, transmission and concentration photometry
  • Up to 10 quantitative standards
  • Four methods for calibration curve: Linear - linear through zero - square - cubic
  • Off-line calculation, re-scaling, curve tracking and peak search
  • Kinetic scanning time course or reacting rate calculations
Delivery information: Supplied with 4 glass cells, 2 quartz cells (not V3000-PC), dust cover, USB cable, 8 GB USB stick, software, 4-positionMore Product Information