VWR® ATC, Handheld Refractometers
Supplier: VWR Collection
VWRI635-0629EA 78.8 SGD
VWRI635-0629 VWRI635-0630 VWRI635-0631 VWRI635-0632 VWRI635-0633 VWRI635-0634 VWRI635-0635 VWRI635-0636 VWRI635-0637 VWRI635-0638 VWRI635-0639
VWR® ATC, Handheld Refractometers
Refractometers Handheld Refractometers
Handheld refractometers with automatic temperature compensation (ATC) for consistent results.
  • ATC at 20 °C between 10 and 30 °C
  • Scales are easy to read and deliver exact results
  • Easy to use and clean