VWR®, pH10 Pen & VWR®ORP15 Pen, pH/°C and ORP/°C Testers

Supplier: VWR Collection
VWRI662-0121EA 331 SGD
VWRI662-0121 VWRI662-0126
VWR®, pH10 Pen & VWR®ORP15 Pen, pH/°C and ORP/°C Testers
Electrochemistry Meters pH and Multi-Parameter Meters
Simple, compact instruments designed for quick accurate results in an economical pen format. Ideal for agriculture, education, fruit and vegetable washing, swimming pools, wastewater, pulp bleaching and aquaculture.

  • Graphic LCD, IP 67 waterproof housing
  • Replaceable electrodes
  • Non volatile memory stores 50 sets of data (pH or absolute/relative ORP, temperature, date and time stamp)
  • GLP functionality - saves and displays last calibration data
  • Automatic temperature compensation; automatic calibration and buffer recognition

Warranty: 1 year (More Product Information
Order Now

SPECIFICATIONS

Model pH10 Pen ORP15 Pen
Environmentally Preferable
pH range 0,00...+14,00 -
pH resolution 0,01 -
pH accuracy ±0,1 -
pH calibration 1-, 2- or 3-point -
mV range - ±1100
mV resolution - 1
mV accuracy - 1 ±1 last significant digit
Temperature range 0,0...+100 °C −10...+100 °C
Temperature resolution 0,1 °C 0,1 °C
Temperature accuracy ±0,3 °C ±0,3 °C
Temperature ATC probe Thermistor, 10 kΩ/25 °C Thermistor, 10 kΩ/25 °C
Display LCD LCD
Power supply 4× LR44 batteries 4× LR44 batteries
Compliance IP 67/GLP IP 67/GLP
Data logging capacity 50 data sets 50 data sets
Weight 0,105 kg (with batteries) 0,105 kg (with batteries)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR