VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
VWRI662-1169EA 268 SGD
VWRI662-1169
VWR® pHenomenal® Electrode Stand
Electrode Holders
Electrode arm moves smoothly in almost any direction, holding up to four electrodes plus one temperature sensor safely and securely in selected positions. Fingertip control raises, lowers, and pivots the holder. The arm articulates at three points while maintaining electrodes in a vertical position.
  • Ideal for multiple readings
  • Accepts all standard electrodes
  • Pivots 360°, extends 53 cm