VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters

Supplier: VWR Collection
VWRI662-1350EA 1370 SGD
VWRI662-1350 VWRI662-2268 VWRI662-2269 VWRI662-2270
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Electrochemistry Meters pH and Multi-Parameter Meters
These economical, robust and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carry case, electrode and temperature sensor with different cable lengths and buffers.

  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic/manual temperature compensation
  • 50 or 250 data sets reviewable memory
  • 1000 hour battery life; auto-shut off function (30 minutes of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display

Additional functions of pH 110 M:
- Extended memory capacityMore Product Information
Order Now

SPECIFICATIONS

Model pH 110 pH 110 M
Environmentally Preferable
pH range −2,00...+16,00 −2,00...+16,00
pH resolution 0,01 0,01
pH accuracy ±0,1% ±2 digits ±0,01% of full scale or ±0,03 pH units
pH calibration 2-point (First point must be a 6,86/7,00 buffer, the second either a 4,00/4,01 or 9,18/10,01) 2-point (First point must be a 6,86/7,00 buffer, the second either a 4,00/4,01 or 9,18/10,01)
Automatic buffer recognition US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
mV range ±1999 ±1999
mV resolution 1 1
mV accuracy ±0,1% ±1 digit ±0,1% ±1 digit
Temperature range −10...+120 °C −10...+120 °C
Temperature resolution 0,1 °C 0,1 °C
Temperature accuracy ±0,3 °C ±2 digits ±0,3 ±1 digit
Temperature compensation Automatic/manual (−10...+120 °C) Automatic/manual (−10...+120 °C)
Power supply One 9 Volt battery One 9 Volt battery
Compliance IP 67 IP 67
Data logging capacity 50 data sets 250 data sets
Outputs - Micro USB
W×D×H 78×37×184 mm 78×37×184 mm
Weight 0,272 kg 0,272 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR