VWR® pHenomenal® pH 1100L/1100LB pH/mV/°C Meter, Bench
Supplier: VWR Collection
VWRI662-1657EA 1830 SGD
VWRI662-1657 VWRI662-1861
VWR® pHenomenal® pH 1100L/1100LB pH/mV/°C Meter, Bench
Electrochemistry Meters pH and Multi-Parameter Meters
This user friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise measurements. Complete with USB output for connection to a computer. GLP compliant.
  • Large graphic LCD with continuous LED backlight showing both pH/mV and temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets
  • Automatic power shut-off: Adjustable from 10 min to 24 h
  • Automatic buffer recognition of DIN and NIST buffers (1,68/4,00/6,86/9,18/12,54
More Product Information