VWR® pHenomenal® IS 2100 L pH/mV/°C/Ion Meter, Bench
Supplier: VWR Collection
VWRI662-1658EA 4200 SGD
VWRI662-1658
VWR® pHenomenal® IS 2100 L pH/mV/°C/Ion Meter, Bench
Electrochemistry Meters pH and Multi-Parameter Meters
This user friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-parameter measurements. Complete with USB output for connection to a computer. GLP compliant.
  • Large graphic LCD with continuous LED backlight shows both pH/mV and temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets
  • Automatic power shut-off: Adjustable from 10 min to 24 h
  • Automatic buffer recognition of DIN and NIST buffers (1,68/4,00/6,86/9
More Product Information