VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR Collection
VWRI662-1350EA 1370 SGD
VWRI662-1350 VWRI662-2268 VWRI662-2269 VWRI662-2270
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Electrochemistry Meters pH and Multi-Parameter Meters
These economical, robust and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carry case, electrode and temperature sensor with different cable lengths and buffers.
  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic/manual temperature compensation
  • 50 or 250 data sets reviewable memory
  • 1000 hour battery life; auto-shut off function (30 minutes of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display
Additional functions of pH 110 M:
- Extended memory capacityMore Product Information