VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
VWRI662-2283EA 349 SGD
VWRI662-2283
VWR® PH 20 pH Meter, Handheld
Electrochemistry Meters pH and Multi-Parameter Meters
This handheld measuring device features an ergonomic design, dust and waterproof housing in accordance with IP 65/67 and an illuminated display. This compact pH meter is a perfect alternative to pH testers.
  • Modern and functional ABS housing
  • 3-line display with backlight
  • HOLD function, min./max. function, low battery indicator
  • Durable, long battery life
  • BNC connection for alternating electrodes
Typical applications include: Aquariums and aquaculture, plant cultivation and agriculture, laboratories, quality assurance, service and food.