VWR® pH 130, pH/mV Meter, Portable
Supplier: VWR Collection
VWRI662-2351EA 1430 SGD
VWRI662-2351
VWR® pH 130, pH/mV Meter, Portable
Electrochemistry Meters pH and Multi-Parameter Meters
These economical, robust, and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carrying case, electrode and temperature sensor with different cable lengths and buffers.
  • Colour, graphic touch screen - enables simultaneous read-out of the pH or mV and temperature value
  • Waterproof housing (IP 66) enables work in difficult conditions
  • Small size and low weight make working in the field easier
  • Calibration of the pH electrode: 1 to 3 points (pH: 4,00 / 7,00 / 10,00)
  • Automatic buffer detection; calibration time reminder
More Product Information