VWR® EC30 Conductivity/TDS/°C Tester, Waterproof
Supplier: VWR Collection
VWRI663-0160EA 466 SGD
VWRI663-0160
VWR® EC30 Conductivity/TDS/°C Tester, Waterproof
Electrochemistry Meters Conductivity Meters
Conductivity, TDS and temperature measurement with simultaneous display of two parameters.
  • Calibration, one or two points
  • Automatic temperature compensation, auto-ranging selection
  • Replaceable conductivity cell
  • IP 67 waterproof casing, low battery indicator
Ordering information: Supplied with 8× LR44 batteries.