VWR® CO 310/CO 310 M, Conductivity Meters, Handheld
Supplier: VWR Collection
VWRI663-0164EA 1970 SGD
VWRI663-0164 VWRI663-0165 VWRI663-0340 VWRI663-0341 VWRI663-0342
VWR® CO 310/CO 310 M, Conductivity Meters, Handheld
Electrochemistry Meters Conductivity Meters
These portable, economical meters can simultaneously measure conductivity, specific conductance, salinity, TDS (Total Dissolved Solids) and temperature.
  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic or manual temperature compensation
  • 50 or 250 data sets reviewable memory
  • 500 (CO 310 M) or 1000 (CO 310) hour battery life; auto-shut off function (30 min of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display
Additional functions of CO 310 M:
- Extended memory capacity (250 data sets)
- Waterproof USBMore Product Information