VWR® pHenomenal® OX 4110 H Dissolved Oxygen Meter, Handheld, with Optical Oxygen Probe
Supplier: VWR Collection
VWRI664-0177EA 4210 SGD
VWRI664-0177
VWR® pHenomenal® OX 4110 H Dissolved Oxygen Meter, Handheld, with Optical Oxygen Probe
Electrochemistry Meters Oxygen Meters
This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise optical oxygen measurements in field applications. Complete with waterproof USB output for connection to a computer. GLP compliant.
  • Large graphic LCD with continuous LED backlight shows both oxygen and temperature values simultaneously
  • Data logging function selectable between 1 min and 1 h
  • 5000 data set memory capacity with automatic storing; additional 200 data set memory for manual storage
  • Automatic power shut-off: Adjustable from 10
More Product Information