VWR® pHenomenal® OPOX 11-3 IDP Optical Dissolved Oxygen Sensor
Supplier: VWR Collection
VWRI664-0182EA 3020 SGD
VWRI664-0182
VWR® pHenomenal® OPOX 11-3 IDP Optical Dissolved Oxygen Sensor
Electrodes Dissolved Oxygen Electrodes
This IDP (Intelligent Digital Probe) dissolved oxygen/temperature sensor can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
  • Post moders type of oxygen measurement: No chemicals, no electrolytes
  • Calibration data (serial number, calibration status) is stored in the electrode/probe, transmitted to the meter and displayed
  • Proven reliability and accuracy
  • Correct measurement results due to direct signal processing in sensor and digital transmission of values
  • All parameters are easily measured with cable lengths of up to 100 metres
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable