VWR® DO 210/DO 210 M, Dissolved Oxygen Meters, Handheld
Supplier: VWR Collection
VWRI664-0081EA 2270 SGD
VWRI664-0081 VWRI664-0082 VWRI664-0228 VWRI664-0229 VWRI664-0230
VWR® DO 210/DO 210 M, Dissolved Oxygen Meters, Handheld
Electrochemistry Meters Oxygen Meters
These precise instruments measure dissolved oxygen in % and ppm (mg/l) and temperature. A built-in microprocessor stores, calculates, and compensates for all parameters related to DO determinations.
  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic temperature compensation
  • 50 or 250 data sets reviewable memory
  • 1000 hour (DO 210) or 500 hour (DO 210 M) battery life; auto-shut off function (after 30 minutes of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display
Additional functions of DO 210 M:
-More Product Information