VWR® DO-230, Portable Meter Set, Dissolved Oxygen
Supplier: VWR Collection
VWRI664-0236EA 1750 SGD
VWRI664-0236
VWR® DO-230, Portable Meter Set, Dissolved Oxygen
Electrochemistry Meters Oxygen Meters
These precise instruments measure dissolved oxygen in % and ppm (mg/L) and temperature. A built-in microprocessor stores, calculates and compensates for all parameters related to DO determinations.
  • Colour, graphic touch screen - enables simultaneous read-out of the oxygen and temperature value
  • Low weight and small size make working in the field easier
  • Easy to use, accurate galvanic oxygen sensor with 1 m cable
  • When properly maintained, the sensor may work efficiently for several years
  • 1- or 2-point oxygen sensor calibration (at 0% and 100% or in the air 0%
More Product Information