VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld

Supplier: VWR Collection
VWRI665-0494EA 2330 SGD
VWRI665-0494 VWRI665-0495 VWRI665-0496
VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld
Electrochemistry Meters pH and Multi-Parameter Meters
These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter and no need to adjust the meter’s settings). The measured data is complete with serial number, settings and calibration data.

  • One universal measuring channel
  • Digital sensor recognition: The measured parameter and settings are recognised automatically
  • Suitable for Intelligent Digital Probes (IDP)
  • Suitable for measurements
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range 0 - 14 0 - 14
pH resolution 0,1/0,01/0,001 0,1/0,01/0,001
pH accuracy ±0,1, ±0,01, ±0,005 ±0,1, ±0,01, ±0,005
mV range ±2500; ±1200 ±2500; ±1200
mV resolution 0,1; 1 0,1; 1
mV accuracy ±0,3; ±1 ±0,3; ±1
Conductivity range 0,01 µS/cm - 1000 mS/cm (depending on sensor) 0,01 µS/cm - 1000 mS/cm (depending on sensor)
Conductivity accuracy ±0,5% of value ±0,5% of value
Conductivity cell constant Automatic Automatic
Resistivity range 1 Ωcm - 20 MΩcm (depending on sensor) 1 Ωcm - 20 MΩcm (depending on sensor)
Salinity range 0 - 70 0 - 70
TDS range 0 mg/L - 199,9 g/L 0 mg/L - 199,9 g/L
DO concentration 0,00 - 20,00 / 0,0 - 50,0 mg/L 0,00 - 20,00 / 0,0 - 50,0 mg/L
DO saturation 0,0 - 200,0; 0 - 500 % 0,0 - 200,0; 0 - 500 %
Temperature range Auto: −5...+105 °C Auto: −5...+105 °C
Temperature resolution 0,1 °C 0,1 °C
Temperature accuracy ±0,2 K ±0,2 K
Display Backlit B/W graphic LCD Blacklit B/W graphic LCD
Data logging - log function Manual/time controlled Manual/time controlled
Data logging capacity Up to 4500 data sets in automatic Up to 4500 data sets in automatic
Memory 4500 points 4500 points
IP rating IP 67 IP 67
Connections 1 measurement input 1 measurement input
Reference temperature 20/25 °C 20/25 °C
Interfaces Mini USB-B (USB 1.1) for data downloads Mini USB-B (USB 1.1) for data downloads
Parameters pH, mV, temperature, conductivity, dissolved oxygen, turbidity pH, mV, temperature, conductivity, dissolved oxygen, turbidity
Calibration Anycal 5-point customer buffer calibration Anycal 5-point customer buffer calibration
Calibration points 1 to 5 (automatic buffer recognition) 1 to 5 (automatic buffer recognition)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR