VWR® pHenomenal® MD 8000 L Bench Top Digital Multi-Parameter Instruments

Supplier: VWR Collection
VWRI665-0499EA 3070 SGD
VWRI665-0499 VWRI665-0498
VWR® pHenomenal® MD 8000 L Bench Top Digital Multi-Parameter Instruments
Electrochemistry Meters pH and Multi-Parameter Meters
These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter, and no need to adjust the meter's settings). The measured data is complete with serial number, settings and calibration data.

  • One universal measuring channel
  • Digital sensor recognition: Measured parameter and settings are recognised automatically
  • Suitable for Intelligent Digital Probes (IDP)
  • Suitable for measurements with
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range 0 - 14
pH resolution 0,1/0,01/0,001
pH accuracy ±0,1; ±0,01; ±0,005
mV range ±2500; ±1200
mV resolution 0,1; 1
mV accuracy ±0,3; ±1
Conductivity range 0,01 µS/cm - 1000 mS/cm (depending on sensor)
Conductivity accuracy ±0,5% of value
Conductivity cell constant Automatic
Resistivity range 1 Ωcm - 20 MΩcm (depending on sensor)
Salinity range 0 - 70
TDS range 0 mg/L - 199,9 g/L
DO concentration 0,00 - 20,00 / 0,0 - 50,0 mg/L
DO saturation 0,0 - 200,0; 0 - 500 %
Temperature range Auto: −5...+105 °C
Temperature resolution 0,1 °C
Temperature accuracy ±0,2 K
Display Blacklit B/W graphic
Power supply 230 V with power supply or 4× 1,5 V AA (supplied) or via USB port
Data logging - log function Yes
Data logging capacity Up to 4500 data sets in automatic
IP rating IP 43
Connections 1 measurement input
Reference temperature 20 / 25 °C
Interfaces Mini USB-B (USB 1.1) for data downloads
Parameters pH, mV, temperature, conductivity, dissolved oxygen, turbidity
Calibration Anycal 5-point customer buffer calibration
Max. 10 calibration storage
1 - 999 days calibration timer
Calibration points 1 - 5 (automatic buffer recognition)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR