VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen

Supplier: VWR Collection
VWRI665-0561EA 2570 SGD
VWRI665-0561
VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen
Electrochemistry Meters pH and Multi-Parameter Meters
User friendly instrument distinguished by a large a 12,7 cm colour graphic touch screen. Can be used for accurate measurements of pH, mV, Redox potential (ORP), conductivity, salinity, TDS, resistivity and temperature. This pH meter enables simultaneous measurements displaying and storing of three functions (pH, conductivity and temperature).

  • Large five colour graphic touch screen showing pH/mV, conductivity and temperature value simultaneously
  • 1- to 3-point calibration with automatic detection of buffer solutions and NIST standard solutions
  • Storing
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range –6,000...+20,000
pH resolution 0,001 / 0,01 pH
pH accuracy ±0,002 pH
pH calibration Automatic, in 1 / 3 points
mV range −2000,0 …+2000,0 mV
mV resolution 0,1 mV / 1 mV
mV accuracy ±0,1 mV
Conductivity range 0,000 − 19,999 mS/cm; 20,00 − 199,99 mS/cm; 200,0 − 1999,9 mS/cm; 2,000 − 19,999 mS/cm; 20,00 − 199,99 mS/cm; 200,0 − 1000,0 mS/cm
Conductivity resolution 0,001 / 0,01 mS/cm; 0,01 / 0,1 mS/cm; 0,1 / 1 mS/cm; 0,001 / 0,01 mS/cm; 0,01 / 0,1 mS/cm; 0,1 / 1 mS/cm
Conductivity accuracy ±0,1%; ±0,1%; ±0,1%; ±0,1%; ±0,25%; ±0,25%
Conductivity calibration 1] By entering the K constant of the cell;
2] With use of max. 3-point calibration
Resistivity range 1,000 - 200 MΩ/cm
Resistivity accuracy 2,0%
TDS range 0 - 1000 g/L
TDS accuracy 1,0%
Temperature range −50,0 …+200,0 °C
Temperature resolution 0,1 °C
Temperature accuracy ±0,1 °C
Temperature compensation Auto/manual
Display 12,7 cm colour graphic touch screen
Data logging - log function Single collection and series of measurements
Data logging capacity 500
Connections BNC for pH/mV banana for temperature
Outputs USB

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR