VWR® shiroGEL Mini PAGE Electroblotting System
Supplier: VWR Collection
VWRI700-0291EA 1980 SGD
VWRI700-0291
VWR® shiroGEL Mini PAGE Electroblotting System
Blotting Apparatus
The shiroGEL Mini PAGE electroblotting system allows quick and easy blotting of three gels simultaneously. Gels fit snugly in blotting cassettes with transfer membranes, which are then placed into the blotting insert. Coiled electrodes within the insert generate a high intensity current, allowing standard transfer to take place in approximately one hour.
  • Blots up to three gels simultaneously
  • Standard transfer in approximately one hour
Delivery information: Complete system includes buffer tank with power leads and cooling pack; blotting insert; 3More Product Information