VWR® shiroGEL Modular mini vertical PAGE systems
Supplier: VWR Collection
VWRI700-0292EA 1940 SGD
VWRI700-0292
VWR® shiroGEL Modular mini vertical PAGE systems
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The shiroGEL mini vertical PAGE system is modular, allowing PAGE and electroblotting to be carried out in the same tank, simply by changing the inserts. The system features inserts for each application, and a common buffer tank in which to run them. Each insert has its own electrode assembly, which connects to the lid of the buffer tank. For safety, the power leads connect directly to the lid. When the lid is removed, power is disconnected from the system. The inserts also have a small pad that fits into position in the lid. This ensures that the lid is always properly placed and aids in itsMore Product Information