VWR® PerfectBlue Mini S, Gel System

Supplier: VWR Collection
VWRI700-0741EA 1440 SGD
VWRI700-0741
VWR® PerfectBlue Mini S, Gel System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The smallest of all the PerfectBlue electrophoresis systems, the Mini S can be used for up to 24 samples enabling short and rapid sample separation. This model includes all the features of the larger models, including direct chamber gel casting and embedded platinum electrodes, and is supplied ready to use with lid, power leads, two combs and a gel tray with an easy to read ruler.

Gel size (W×L): 70×80 mm

Accessories information: * Based on an agarose gel of approximately 5 mm thickness

Ordering information: System is supplied with 2 combs (6 and 10 teeth, 1,5 mm thickness) as standard.
Order Now

SPECIFICATIONS

Model Mini S
Environmentally Preferable
Gel size W×L 70×80 mm
Gel volume ~30 ml
Buffer volume ~400 ml
Run time ≥30 h
Unit W×L×H 120×160×95 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR