VWR® PerfectBlue Mini L, Gel System
Supplier: VWR Collection
VWRI700-0777EA 1780 SGD
VWRI700-0777
VWR® PerfectBlue Mini L, Gel System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Capable of running 12×14 cm gels, the largest of the 'Mini' range still offers the convenience of direct gel casting in the tray, in an easy to manage product size for reliable, high quality electrophoresis. This system can be supplied with a choice of gel trays suitable for 2, 4 or 12 combs, and an expansive range of standard and microtitre combs, making this versatile system appropriate for both routine and small separation electrophoresis.
*Based on an agarose gel of approximately 5 mm thickness
Ordering information: System is supplied with gasketed UV transmissible (UVT) gel tray (More Product Information