VWR® PerfectBlue Mini L Revolution, Gel System
Supplier: VWR Collection
VWRI700-0799EA 2700 SGD
VWRI700-0799
VWR® PerfectBlue Mini L Revolution, Gel System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The 'Revolution' series includes a buffer circulation system, powered by the electrodes themselves. Hydrogen bubbles produced at the cathode pass up a diagonally positioned pipe connected to the two sides of the tank producing a gentle circulation of the buffer. The result is no pH and ionic gradient – without need of mechanical pumps.
*Based on an agarose gel of approximately 5 mm thickness
Ordering information: System is supplied with 2 combs (12 and 20 teeth, 1,5 mm thickness) as standard.