VWR® PerfectBlue Maxi M, Gel system
Supplier: VWR Collection
VWRI700-0880EA 2790 SGD
VWRI700-0880
VWR® PerfectBlue Maxi M, Gel system
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Wider than the Maxi S Plus, the Maxi M model can accommodate up to 168 samples but still with an economical footprint.
*Based on an agarose gel of approximately 5 mm thickness.
Ordering information: Supplied with 3 combs (1,5 mm thickness, 16, 24, 36 teeth), lid, gel tray, 3-point levelling platform, end gates, tank and cables.