VWR® PerfectBlue Maxi ExW, Wide format gel system
Supplier: VWR Collection
VWRI700-0910EA 3110 SGD
VWRI700-0910
VWR® PerfectBlue Maxi ExW, Wide format gel system
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
This 'extra wide' Maxi tank was developed especially for large sample numbers and rapid separation times. Accommodating up to 10 microtitre combs, the Maxi ExW can be used to separate up to 500 samples and facilitates error-free loading with multichannel pipettes. By optimising the position of the embedded platinum electrode and the use of the three-point levelling platform, with in-built bubble level, the Maxi ExW provides highly reproducible and dependable sample separation, free from 'smiling' effects.
The tray has 10 comb positions accomodating two different comb thicknesses. The trayMore Product Information