VWR® PerfectBlue Maxi ExW 'Revolution', Wide format gel system
Supplier: VWR Collection
VWRI700-0912EA 3940 SGD
VWRI700-0912
VWR® PerfectBlue Maxi ExW 'Revolution', Wide format gel system
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The Maxi ExW 'Revolution' offers built-in buffer recirculation - without the need for pumps or additional tubing. The Maxi ExW 'Revolution' offers the same high throughput, rapid separation as the Maxi ExW model but, with the position of the cathode and a rising tube, the system uses hydrogen bubbles to slowly mix the buffer. This system thereby avoids the build-up of pH or ionic gradients for electrophoresis of maximum quality and stability.
The Maxi ExW 'Revolution' system offers 10 microtitre comb positions with a maximum capacity for 500 samples. The optional wall combs or castingMore Product Information