VWR® M40, Transport Swab
Supplier: VWR Collection
VWRI710-0442EA 75.5 SGD
VWRI710-0442
VWR® M40, Transport Swab
Swabs Transport Swabs
Transport swab with plastic shaft, synthetic tip, and Amies agar gel.
  • Compliant with CLSI (formerly NCCLS) standard M40-A at 21 and 4 °C
  • Without charcoal
Helps to improve recovery and survival of fastidious microorganisms from the throat, vagina, wounds and skin swabs.
Certifications: CLSI (formerly NCCLS).
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904