VWR® SP100 Automatic Straw Pipette, Fixed Volume
Supplier: VWR Collection
VWRI710-1021EA 729 SGD
VWRI710-1021
VWR® SP100 Automatic Straw Pipette, Fixed Volume
Pipettes Straw Pipettes
Air displacement pipette with two pre-calibrated steps allows subsequent pipetting of 1 and 0,1 ml of the same liquid. Ideal alternative to graduated glass pipettes when performing serial 1:10 dilutions in bacteriology.
  • Smooth activation, excellent ergonomics, easy maintenance, cleaning and disinfection
  • Two pre-calibrated fixed volumes – no setting required
  • Both volumes calibrated independently
  • Interchangeable PE nozzle protection filter
  • Justip™ system for tip ejector height adjustment
  • Autoclavable
Caution: *The product(s) below are outside the scopeMore Product Information