VWR® Auto lite, Automatic Colony Counters
Supplier: VWR Collection
VWRI710-2072EA 15890 SGD
VWRI710-2072
VWR® Auto lite, Automatic Colony Counters
Colony Counters
Auto lite is the latest affordable, automated colony counting device. It is available in one configuration and features simple to use software that greatly improves efficiency, reliability and productivity. Auto lite software uses a powerful algorithm for colony counting on pour and spiral plates. A colony separator automatically splits touching colonies to further improve the accuracy of the counting process.
  • Low cost entry into electronic colony counting, pour and spiral
  • Results and plate images are saved to a PC enabling traceability, consistency
More Product Information