VWR® Q Path®, Cytology Funnel
Supplier: VWR Collection
VWRI720-1999EA 687 SGD
VWRI720-1999
VWR® Q Path®, Cytology Funnel
Funnels Cytology Funnels
Single-use plastic chamber used in cytocentrifugation for urine, gynaecological and non gynaecological specimens, also designed to perform cell suspension transfers on a slide with low risk of contamination.
  • Two apertures with attached filter - large absorption capacity
  • Can resist water and alcohol for 24 H
  • Speed of absorption is ±1 ml in three minutes
Order Now
SPECIFICATIONS
Environmentally Preferable