VWR® Q Path® MacroStar II, Cassettes, in Sleeve
Supplier: VWR Collection
VWRI720-2052EA 618 SGD
VWRI720-2052 VWRI720-2054 VWRI720-2055 VWRI720-2056 VWRI720-2057 VWRI720-2058 VWRI720-2051 VWRI720-2053 VWRI720-2050
VWR® Q Path® MacroStar II, Cassettes, in Sleeve
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes in pre-filled sleeves for Thermo and RA Lamb cassette printers. 50 dispensers with 80 cassettes and 2×2000 lids.
  • Large opening for the filtration of fluids
  • Hinge and separate lid
  • With 45° writing angle
Order Now
SPECIFICATIONS
Environmentally Preferable