VWR® Q Path® MacroStar V, Cassettes, in Racks
Supplier: VWR Collection
VWRI720-2163EA 286 SGD
VWRI720-2163 VWRI720-2164 VWRI720-2165 VWRI720-2166 VWRI720-2167 VWRI720-2168 VWRI720-2169 VWRI720-2170 VWRI720-2099
VWR® Q Path® MacroStar V, Cassettes, in Racks
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Cassettes with welded disposable lid, supplied in racks for Leica and Sakura cassette printers. 2000 cassettes packed 40/rack.
  • Easy to use cassette with large opening
  • 45° writing area at the front and on both sides
  • Cassettes with ridges, which makes it easier to be opened after impregnation
Order Now
SPECIFICATIONS
Environmentally Preferable