VWR® Q Path® MicroStar V Cassettes
Supplier: VWR Collection
VWRI720-2194EA 174 SGD
VWRI720-2194 VWRI720-2195 VWRI720-2199 VWRI720-2197 VWRI720-2192 VWRI720-2193 VWRI720-2196 VWRI720-2198 VWRI720-2205
VWR® Q Path® MicroStar V Cassettes
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with welded disposable lid, supplied in dispensers for 500 cassettes. Packed 3 boxes with 500 cassettes.
  • Easy to use cassette with small opening
  • Writing at the front and on both sides
  • Cassettes with ridges, which makes it easier to be opened after impregnation
Order Now
SPECIFICATIONS
Environmentally Preferable