VWR® Q Path® MacroStar V, Cassettes
Supplier: VWR Collection
VWRI720-2219EA 170 SGD
VWRI720-2219 VWRI720-2220 VWRI720-2215 VWRI720-2221 VWRI720-2222 VWRI720-2223 VWRI720-2216 VWRI720-2218 VWRI720-2217
VWR® Q Path® MacroStar V, Cassettes
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with welded disposable lid, supplied in dispensers for 500 cassettes. Packed in three boxes of 500 cassettes.
  • Easy to use cassette
  • 45° writing area at the front and on both sides
  • Cassettes with ridges, which makes it easier to be opened after impregnation
Order Now
SPECIFICATIONS
Environmentally Preferable