VWR® Q Path® MacroStar II, Cassettes, in Dispenser
Supplier: VWR Collection
VWRI720-2391EA 334 SGD
VWRI720-2391 VWRI720-2392 VWRI720-2393 VWRI720-2394 VWRI720-2395 VWRI720-2396 VWRI720-2397 VWRI720-2398 VWRI720-2399
VWR® Q Path® MacroStar II, Cassettes, in Dispenser
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with tape (= rack) for Leica and Sakura cassette printers. Rack of 40 cassettes.
  • Large opening for the filtration of fluids
  • Hinge and separate lid
  • 45° writing angle
Order Now
SPECIFICATIONS
Environmentally Preferable