VWR®, Prepared Slides: Phanerogams
Supplier: VWR Collection
VWRI720-2926EA 194 SGD
VWRI720-2926
VWR®, Prepared Slides: Phanerogams
Slides Prepared Slides
Prepared slides of seed bearing plants (phanerogams), including.
  • Pine tree leaf (c.s.)
  • Pine tree, 1 year old stem (c.s.)
  • Pine tree, 10 year old stem (c.s.)
  • Pine tree, spherical male fruit (l.s.)
  • Pine tree, spherical female fruit (l.s.)
  • Pine tree, pollen (w.b.)
  • Buttercup root (c.s.)
  • Corn (mais) root (c.s.)
  • Mitosis, onion tips (s.)
  • Geranium stem (c.s.)
  • Marchantia antheridiophore (l.s.)
  • Marchantia archegoniophore (l.s.)
  • Sunflower, young stem (c.s.)
  • Tilia (linden tree), 1 year old stem (c.s.)
  • Tilia (linden tree), 3 year old stem (c.s.)
  • Lily leaf (c.s.)
  • Fig
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable