VWR®, Prepared Slides for Zoology: Vertebrates and Mammals
Supplier: VWR Collection
VWRI720-2928EA 237 SGD
VWRI720-2928
VWR®, Prepared Slides for Zoology: Vertebrates and Mammals
Slides Prepared Slides
Prepared slides for zoology i.e. vertebrates and mammals including.
  • Frog skin (c.s.), small intestine (c.s.), blood smear, testis (s.), liver (s.) and heart (s.)
  • Rabbit liver (s.), testis (s.), vertebral column (c.s.), motor nerve endings (w.b.) and ovary (s.)
  • Compact bone tissue
  • Stomach walls (s.)
  • Mammal small intestine (c.s.), large intestine (c.s.), pancreas, spleen (s.), gall bladder (s.), lung (s.), artery and vein (c.s.), kidney (l.s.) and eyeball (l.s.)
  • Human sperm smear, skin (s.) through sweat gland and chromosome in blood, male and female