VWR® shiroGEL horizontal 96, Horizontal Electrophoresis Apparatus
Supplier: VWR Collection
VWRI730-0340EA 1660 SGD
VWRI730-0340
VWR® shiroGEL horizontal 96, Horizontal Electrophoresis Apparatus
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
This 96-well format horizontal gel electrophoresis unit is compatible with 8-channel multichannel pipettes, and matches up with a standard 96-well plate configuration.
  • Average run time is 15 to 30 min per gel
  • Layout of wells allows a run length of 18 mm per sample, providing ideal separation